3-Bromo-5-iodo-1H-indazole 95%
Supplier: Ambeed
|
Warning
|
A143162-1G
A143162-250MG
A143162-5G
76823-520EA
153.84
USD
76823-520
76823-522
76823-524
3-Bromo-5-iodo-1H-indazole 95%
3-Bromo-5-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄BrIN₂ |
MDL Number:
MFCD07781644 CAS Number: 885519-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...