D-Lysine methyl ester dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Lys-OMe.2HCl, (R)-Methyl 2,6-diaminohexanoate dihydrochloride

A142669-10G A142669-250MG A142669-25G A142669-5G
76823-510EA 30 USD
76823-510 76823-512 76823-514 76823-516 76823-518
D-Lysine methyl ester dihydrochloride 97%
D-Lysine methyl ester dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O₂
MDL Number: MFCD00039067
CAS Number: 67396-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR