4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde, 2-Formylthiophene-4-boronic acid pinacol ester

A141600-10G A141600-1G A141600-250MG A141600-25G A141600-5G
76823-476EA 231.28 USD
76823-476 76823-478 76823-480 76823-482 76823-484
4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde 98%
4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₃S
MW: 238.111 g/mol
MDL Number: MFCD12756464
CAS Number: 881381-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR