3-Bromo-5-(methylsulfonyl)aniline 98%

Supplier: Ambeed
Warning

A141349-1G A141349-250MG
76823-454EA 203.7 USD
76823-454 76823-456
3-Bromo-5-(methylsulfonyl)aniline 98%
3-Bromo-5-(methylsulfonyl)aniline

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂S
MW: 250.113 g/mol
MDL Number: MFCD17392822
CAS Number: 62606-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR