Methyl 3-chloro-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A140982-1G A140982-250MG A140982-5G A140982-25G
76823-438EA 14.86 USD
76823-438 76823-440 76823-442 77683-229
Methyl 3-chloro-2-nitrobenzoate 98%
Methyl 3-chloro-2-nitrobenzoate

Precaución: For research use only.

Formula: C₈H₆ClNO₄
MW: 215.591 g/mol
MDL Number: MFCD04038813
CAS Number: 42087-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR