8-Bromo-3,4-dihydrobenzo[b]oxepin-5(2H)-one 97%

Supplier: Ambeed
Warning

A140747-100MG A140747-1G A140747-250MG A140747-5G
76823-430EA 14 USD
76823-430 76823-432 76823-434 76823-436
8-Bromo-3,4-dihydrobenzo[b]oxepin-5(2H)-one 97%
8-Bromo-3,4-dihydrobenzo[b]oxepin-5(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MDL Number: MFCD09040560
CAS Number: 141106-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR