3-Fluoro-5-iodobenzotrifluoride 95%

Supplier: Ambeed
Warning

A140659-1G A140659-250MG
76823-420EA 252 USD
76823-420 76823-422
3-Fluoro-5-iodobenzotrifluoride 95%
3-Fluoro-5-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄I
MDL Number: MFCD09800718
CAS Number: 1027513-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR