2-(Ethylamino)-5-pyrimidineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A140405-100MG A140405-250MG A140405-5G
76823-398EA 25 USD
76823-398 76823-400 76823-402
2-(Ethylamino)-5-pyrimidineboronic acid pinacol ester 95%
2-(Ethylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₂₀BN₃O₂
MW: 249.1200 g/mol
MDL Number: MFCD12546554
CAS Number: 1218791-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR