1-(2-Iodoethyl)-4-octylbenzene 95%

Supplier: Ambeed
Warning

A139926-25G A139926-5G
76823-386EA 169.75 USD
76823-386 76823-388
1-(2-Iodoethyl)-4-octylbenzene 95%
1-(2-Iodoethyl)-4-octylbenzene

Precaución: for Research Use Only

Formula: C₁₆H₂₅I
MW: 344.27 g/mol
MDL Number: MFCD09753530
CAS Number: 162358-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR