4-Bromo-7-hydroxy-2,3-dihydro-1H-inden-1-one 95%
Supplier: Ambeed
|
Warning
|
A139299-100MG
A139299-1G
A139299-5G
76823-360EA
21.22
USD
76823-360
76823-362
76823-364
4-Bromo-7-hydroxy-2,3-dihydro-1H-inden-1-one 95%
4-Bromo-7-hydroxy-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only
Formula:
C₉H₇BrO₂ MW: 227.06 g/mol |
MDL Number:
MFCD00463881 CAS Number: 81945-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...