2-Iodo-5-methoxyphenol 97%

Supplier: Ambeed
Warning

A139160-100MG A139160-250MG A139160-5G A139160-1G
76823-352EA 23 USD
76823-352 76823-354 76823-356 77885-175
2-Iodo-5-methoxyphenol 97%
2-Iodo-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇IO₂
MDL Number: MFCD12963849
CAS Number: 41046-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR