Methyl 6-amino-1H-indazole-7-carboxylate 98%

Supplier: Ambeed
Warning

A138328-250MG A138328-5G
76823-344EA 44 USD
76823-344 77566-780
Methyl 6-amino-1H-indazole-7-carboxylate 98%
Methyl 6-amino-1H-indazole-7-carboxylate

Precaución: For research use only.

Formula: C₉H₉N₃O₂
MW: 191.19 g/mol
MDL Number: MFCD20691477
CAS Number: 73907-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR