3'-Bromo-5'-fluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Bromo-5-fluorophenyl)ethanone

A138144-100MG A138144-1G A138144-250MG A138144-25G A138144-5G
76823-318EA 5.31 USD
76823-318 76823-320 76823-322 76823-324 76823-326
3'-Bromo-5'-fluoroacetophenone 97%
3'-Bromo-5'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MW: 217.04 g/mol
MDL Number: MFCD11847746
CAS Number: 105515-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR