5-Bromo-4-methylpicolinaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-methylpyridine-2-carbaldehyde

A138091-100MG A138091-1G A138091-250MG A138091-5G
76823-304EA 26.53 USD
76823-304 76823-306 76823-308 76823-310
5-Bromo-4-methylpicolinaldehyde 97%
5-Bromo-4-methylpicolinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 200.033 g/mol
MDL Number: MFCD07375095
CAS Number: 886364-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR