(R)-4-Benzyl-3-(chloromethyl)morpholine 97%

Supplier: Ambeed
Warning

A137893-100MG A137893-1G A137893-250MG
76823-298EA 10.61 USD
76823-298 76823-300 76823-302
(R)-4-Benzyl-3-(chloromethyl)morpholine 97%
(R)-4-Benzyl-3-(chloromethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO
MW: 225.72 g/mol
MDL Number: MFCD10687125
CAS Number: 1217697-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR