6-Fluoro-1H-indol-4-amine 97%

Supplier: Ambeed
Warning

A137779-1G A137779-100MG
76823-296EA 268.41 USD
76823-296 77683-209
6-Fluoro-1H-indol-4-amine 97%
6-Fluoro-1H-indol-4-amine

Precaución: For research use only.

Formula: C₈H₇FN₂
MW: 150.153 g/mol
MDL Number: MFCD07357294
CAS Number: 885518-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR