Methyl 4,6-dimethylnicotinate 95%

Supplier: Ambeed
Warning

A137748-1G A137748-250MG
76823-292EA 463.62 USD
76823-292 76823-294
Methyl 4,6-dimethylnicotinate 95%
Methyl 4,6-dimethylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD18256178
CAS Number: 69971-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR