3-Bromo-5-chlorophenylboronic acid 95%

Supplier: Ambeed
Warning

A137052-10G A137052-1G A137052-250MG A137052-25G A137052-5G A137052-100G
76823-266EA 19 USD
76823-266 76823-268 76823-270 76823-272 76823-274 77885-165
3-Bromo-5-chlorophenylboronic acid 95%
3-Bromo-5-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MDL Number: MFCD13189469
CAS Number: 1186403-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR