2-(4-Aminophenyl)oxazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Oxazol-2-yl)aniline

A136895-100MG A136895-1G A136895-250MG
76823-258EA 51.99 USD
76823-258 76823-260 76823-262
2-(4-Aminophenyl)oxazole 98%
2-(4-Aminophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.173 g/mol
MDL Number: MFCD11040381
CAS Number: 62882-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR