4-Methylpyridazine-3,6-dione 95%

Supplier: Ambeed
Warning

A136815-100MG A136815-1G A136815-250MG
76823-252EA 28 USD
76823-252 76823-254 76823-256
4-Methylpyridazine-3,6-dione 95%
4-Methylpyridazine-3,6-dione

Precaución: for Research Use Only

Formula: C₅H₄N₂O₂
MDL Number: MFCD22056209
CAS Number: 73268-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR