7-Bromoquinolin-4(1H)-one 97%

Supplier: Ambeed
Warning

A136717-1G A136717-250MG
76823-242EA 81.69 USD
76823-242 76823-244
7-Bromoquinolin-4(1H)-one 97%
7-Bromoquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.05 g/mol
MDL Number: MFCD19703293
CAS Number: 956268-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR