1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride 95%

Supplier: Ambeed
Warning

Synonyms: 5,12-Dibromoanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone

A136432-1G A136432-250MG A136432-25G A136432-5G
76823-234EA 28.65 USD
76823-234 76823-236 76823-238 76823-240
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride 95%
1,7-Dibromo-3,4,9,10-perylenetetracarboxylic dianhydride

Precaución: for Research Use Only

Formula: C₂₄H₆Br₂O₆
MW: 550.11 g/mol
MDL Number: MFCD11870875
CAS Number: 118129-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR