2-Amino-5-phenylpentanoic acid 97%

Supplier: Ambeed
Warning

A136266-100MG A136266-1G A136266-250MG
76823-222EA 54.11 USD
76823-222 76823-224 76823-226
2-Amino-5-phenylpentanoic acid 97%
2-Amino-5-phenylpentanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.24 g/mol
MDL Number: MFCD02322597
CAS Number: 34993-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR