1-(3-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 97%

Supplier: Ambeed
Warning

A135659-100MG A135659-250MG
76823-210EA 76.39 USD
76823-210 76823-212
1-(3-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 97%
1-(3-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClF₃N
MW: 237.65 g/mol
MDL Number: MFCD09028132
CAS Number: 1108698-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR