2-(Ethylsulfonyl)aniline 95%

Supplier: Ambeed
Warning

A135433-1G A135433-250MG
76823-200EA 220.67 USD
76823-200 76823-202
2-(Ethylsulfonyl)aniline 95%
2-(Ethylsulfonyl)aniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MW: 185.243 g/mol
MDL Number: MFCD09945923
CAS Number: 31596-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR