5-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A135206-1G A135206-250MG
76823-188EA 35.01 USD
76823-188 76823-190 76823-192
5-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid 97%
5-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂O₂
MDL Number: MFCD13193407
CAS Number: 1260798-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR