Dodecyl 2-(dimethylamino)propanoate hydrochloride 95%

Supplier: Ambeed
Warning

A134964-1G A134964-250MG
76823-182EA 121 USD
76823-182 76823-184
Dodecyl 2-(dimethylamino)propanoate hydrochloride 95%
Dodecyl 2-(dimethylamino)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₃₆ClNO₂
MW: 321.92 g/mol
MDL Number: MFCD20040092
CAS Number: 259685-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR