5-Chloro-2-methoxy-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-methoxypyridine-3-boronic acid, (5-Chloro-2-methoxypyridin-3-yl)boronic acid

A134667-1G A134667-250MG A134667-5G
76823-168EA 44.56 USD
76823-168 76823-170 76823-172
5-Chloro-2-methoxy-3-pyridineboronic acid 98%
5-Chloro-2-methoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
MDL Number: MFCD04114579
CAS Number: 943153-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR