4-Methyl-6-(methylthio)pyrimidin-2-ol 98%

Supplier: Ambeed
Warning

A134549-100MG A134549-1G A134549-250MG
76823-162EA 37.14 USD
76823-162 76823-164 76823-166
4-Methyl-6-(methylthio)pyrimidin-2-ol 98%
4-Methyl-6-(methylthio)pyrimidin-2-ol

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MW: 156.21 g/mol
MDL Number: MFCD00090779
CAS Number: 16710-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR