2-Methyl-2-(4-nitrophenyl)propanenitrile 97%

Supplier: Ambeed
Warning

A133922-100MG A133922-1G A133922-250MG A133922-25G A133922-5G
76823-142EA 5.31 USD
76823-142 76823-144 76823-146 76823-148 76823-150
2-Methyl-2-(4-nitrophenyl)propanenitrile 97%
2-Methyl-2-(4-nitrophenyl)propanenitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD00844930
CAS Number: 71825-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR