6-Chloro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A133819-100MG A133819-1G A133819-250MG
76823-136EA 47.75 USD
76823-136 76823-138 76823-140
6-Chloro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%
6-Chloro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉ClN₂O₂S
MW: 292.74 g/mol
MDL Number: MFCD11616448
CAS Number: 896722-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR