Tetraethylene glycol bis(p-toluenesulfonate) 97%

Supplier: Ambeed
Warning

Synonyms: Bis[2-[2-(p-toluenesulphonyloxy)ethoxy]ethyl] Ether, ((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), Bis-Tos-PEG4, Bis[2-(2-tosyloxyethoxy)ethyl] Ether, Tetraethylene Glycol Di-p-tosylate

A133747-100G A133747-10G A133747-1G A133747-5G A133747-25G A133747-500G
76823-128EA 200.52 USD
76823-128 76823-130 76823-132 76823-134 77549-604 77683-187
Tetraethylene glycol bis(p-toluenesulfonate) 97%
Tetraethylene glycol bis(p-toluenesulfonate)

Precaución: For research use only.

Formula: C₂₂H₃₀O₉S₂
MW: 502.59 g/mol
MDL Number: MFCD00075239
CAS Number: 37860-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR