4-Bromo-2-methylindan-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methyl-2,3-dihydro-1H-inden-1-one, 4-Bromo-2-methyl-1-indanone

A133397-1G A133397-5G
76823-118EA 57.29 USD
76823-118 76823-120
4-Bromo-2-methylindan-1-one 95%
4-Bromo-2-methylindan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrO
MW: 225.09 g/mol
MDL Number: MFCD09881709
CAS Number: 174702-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR