CHIR99021 98%

Supplier: Ambeed
Warning

Synonyms: CHIR 99021, 6-((2-((4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino)nicotinonitrile, CHEMBL412142, GSK-3 Inhibitor XVI, AGN-PC-0MWAEX

A133052-10MG A133052-1MG A133052-250MG A133052-5MG A133052-100MG A133052-1G A133052-50MG
76823-110EA 32.89 USD
76823-110 76823-112 76823-114 76823-116 77683-178 77683-179 77683-180
CHIR99021 98%
CHIR99021

Precaución: For research use only.

Formula: C₂₂H₁₈Cl₂N₈
MW: 465.3379 g/mol
MDL Number: MFCD11846251
CAS Number: 252917-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR