2,4-Dimethylbenzo[d]thiazole 98%

Supplier: Ambeed
Warning

A132964-100MG A132964-1G A132964-250MG A132964-5G
76823-102EA 29.71 USD
76823-102 76823-104 76823-106 76823-108
2,4-Dimethylbenzo[d]thiazole 98%
2,4-Dimethylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₉H₉NS
MW: 163.23 g/mol
MDL Number: MFCD03161856
CAS Number: 5262-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR