2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl, 3-Biphenylboronic acid Pinacol ester

A132927-10G A132927-1G A132927-250MG A132927-25G A132927-5G
76823-092EA 58.35 USD
76823-092 76823-094 76823-096 76823-098 76823-100
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-([1,1'-Biphenyl]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₈H₂₁BO₂
MW: 280.17 g/mol
MDL Number: MFCD18452169
CAS Number: 912844-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR