Sodium (4-nitrophenyl)methanesulfonate 98%

Supplier: Ambeed
Warning

A132898-100G A132898-100MG A132898-250MG A132898-25G A132898-5G A132898-1G
76823-080EA 335 USD
76823-080 76823-082 76823-084 76823-086 76823-088 77885-144
Sodium (4-nitrophenyl)methanesulfonate 98%
Sodium (4-nitrophenyl)methanesulfonate

Precaución: for Research Use Only

Formula: C₇H₆NNaO₅S
MDL Number: MFCD02334757
CAS Number: 36639-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR