4-Amino-1-(3-methoxypropyl)piperidine 97%

Supplier: Ambeed
Danger

Synonyms: 1-(3-Methoxypropyl)piperidin-4-amine

A132725-10G A132725-1G A132725-250MG A132725-5G
76823-072EA 170.81 USD
76823-072 76823-074 76823-076 76823-078
4-Amino-1-(3-methoxypropyl)piperidine 97%
4-Amino-1-(3-methoxypropyl)piperidine

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O
MW: 172.27 g/mol
MDL Number: MFCD11104531
CAS Number: 179474-79-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR