2,5-Dibromo-3,4-pyridinediamine 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromopyridine-3,4-diamine

A131486-100MG A131486-1G A131486-250MG A131486-25G A131486-5G A131486-10G
76823-034EA 20.16 USD
76823-034 76823-036 76823-038 76823-040 76823-042 77683-166
2,5-Dibromo-3,4-pyridinediamine 98%
2,5-Dibromo-3,4-pyridinediamine

Precaución: For research use only.

Formula: C₅H₅Br₂N₃
MW: 266.921 g/mol
MDL Number: MFCD15526814
CAS Number: 221241-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR