Methyl 2-Acetylpyridine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-acetylisonicotinate

A131287-10G A131287-1G A131287-250MG A131287-25G A131287-5G A131287-100G
76823-024EA 78.51 USD
76823-024 76823-026 76823-028 76823-030 76823-032 77592-514
Methyl 2-Acetylpyridine-4-carboxylate 98%
Methyl 2-Acetylpyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.173 g/mol
MDL Number: MFCD09032412
CAS Number: 138715-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR