2,6-Dichloro-N-methylpyridin-4-amine 97%

Supplier: Ambeed
Warning

A130367-100MG A130367-1G A130367-250MG
76822-990EA 35.01 USD
76822-990 76822-992 76822-994
2,6-Dichloro-N-methylpyridin-4-amine 97%
2,6-Dichloro-N-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.031 g/mol
MDL Number: MFCD12024871
CAS Number: 175461-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR