5-(Hydroxymethyl)-3-methylisoxazole 97%

Supplier: Ambeed
Warning

Synonyms: (3-Methyl-1,2-oxazol-5-yl)methanol, (3-Methylisoxazol-5-yl)methanol

A130236-100MG A130236-1G A130236-250MG A130236-5G
76822-982EA 67.9 USD
76822-982 76822-984 76822-986 77683-156
5-(Hydroxymethyl)-3-methylisoxazole 97%
5-(Hydroxymethyl)-3-methylisoxazole

Precaución: For research use only.

Formula: C₅H₇NO₂
MW: 113.115 g/mol
MDL Number: MFCD09859210
CAS Number: 14716-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR