DL-Pipecolic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A130065-10G
A130065-1G
A130065-250MG
A130065-5G
76822-960EA
31
USD
76822-960
76822-962
76822-964
76822-966
DL-Pipecolic acid hydrochloride 95%
DL-Pipecolic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₂·ClH |
MDL Number:
MFCD00012771 CAS Number: 15862-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...