5-Chloro-2-methylnicotinonitrile 98%

Supplier: Ambeed
Warning

A130023-250MG A130023-50MG
76822-956EA 135.8 USD
76822-956 76822-958
5-Chloro-2-methylnicotinonitrile 98%
5-Chloro-2-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD18255242
CAS Number: 1256819-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR