2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(6-phenyl-3-pyridyl)-1,3,2-dioxaborolane, 6-Phenylpyridine-3-boronic acid pinacol ester, 6-Phenyl-3-pyridylboronic acid Pinacol ester

A129739-100MG A129739-1G A129739-250MG
76822-950EA 21 USD
76822-950 76822-952 77885-137
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₂
MDL Number: MFCD12032565
CAS Number: 879291-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR