7-Bromoquinoline-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A129317-100MG A129317-1G A129317-250MG A129317-5G
76822-936EA 85.94 USD
76822-936 76822-938 77683-147 77683-148
7-Bromoquinoline-4-carboxylic acid 95%
7-Bromoquinoline-4-carboxylic acid

Precaución: For research use only.

Formula: C₁₀H₆BrNO₂
MW: 252.06 g/mol
MDL Number: MFCD11108751
CAS Number: 31009-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR