Potassium 2-hydroxy-4-methoxybenzoate 97%

Supplier: Ambeed
Warning

A129213-100G A129213-10G A129213-1G A129213-25G A129213-5G
76822-928EA 242.95 USD
76822-928 76822-930 76822-932 76822-934 77683-146
Potassium 2-hydroxy-4-methoxybenzoate 97%
Potassium 2-hydroxy-4-methoxybenzoate

Precaución: For research use only.

Formula: C₈H₇KO₄
MW: 206.23 g/mol
MDL Number: MFCD13194966
CAS Number: 152312-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR