1-(4-Bromobenzoyl)pyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)(pyrrolidin-1-yl)methanone

A129128-100G A129128-1G A129128-250MG A129128-25G A129128-5G
76822-906EA 1031.2 USD
76822-906 76822-908 76822-910 76822-912 76822-914
1-(4-Bromobenzoyl)pyrrolidine 97%
1-(4-Bromobenzoyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MW: 254.13 g/mol
MDL Number: MFCD00595283
CAS Number: 5543-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR