2-Chloro-4-methyl-5-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-methoxy-5-methyl-4-nitrobenzene, 2-Chloro-4-methyl-5-nitrophenyl methyl ether

A129023-100MG A129023-10G A129023-1G A129023-250MG
76822-898EA 11.67 USD
76822-898 76822-900 76822-902 76822-904
2-Chloro-4-methyl-5-nitroanisole 98%
2-Chloro-4-methyl-5-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.607 g/mol
MDL Number: MFCD23703045
CAS Number: 62492-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR