2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene 95%

Supplier: Ambeed
Warning

Synonyms: N-([1,1'-Biphenyl]-4-yl)-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine

A128992-100G A128992-10G A128992-1G A128992-25G A128992-5G
76822-882EA 339 USD
76822-882 76822-884 76822-886 76822-888 76822-890
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene 95%
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene

Precaución: for Research Use Only

Formula: C₃₃H₂₆BrN
MW: 516.48 g/mol
MDL Number: MFCD20040455
CAS Number: 1246562-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR